Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Yeast Lytic Enzyme, from Arthrobacter luteus

Catalog No. ICN15352650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15352650 50 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15352650 Supplier MP Biomedicals™ Supplier No. 0215352650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Activity 20,000units/g

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20,000 U/g
Color Tan
pH 7.5
For Use With (Application) Used to digest the cell wall of active Yeast cells
Content And Storage 0°C
Source Arthrobacter luteus
Quantity 50 kU
Product Type Yeast Lytic Enzyme
Form Lyophilized Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.