Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Yeast Lytic Enzyme

Catalog No. ICN15352610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15352610 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15352610 Supplier MP Biomedicals™ Supplier No. 0215352610
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

From Arthrobacter luteus

Unit Definition: One unit will produce a ΔA800 of 0.001 per minute at pH 7.5 and 25°C, using a suspension of brewers yeast as a substrate in a 3 mL reaction mixture.

This enzyme catalyzes the lysis of yeast cells and hydrolyzes glucose polymers with β-1,3-linkages (e.g., glucan). It is also useful for preparing yeast spheroplasts.

Zymolyase
Lyophilized powder
Activity: 20,000 units/g

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20,000 U/g
Color Tan
pH 7.5
For Use With (Application) Digest the cell wall of active Yeast cells
Content And Storage 0°C
Source Arthrobacter luteus
Quantity 10 kU
Product Type Yeast Lytic Enzyme
Form Lyophilized Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.