Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YEP Broth, Dehydrated Media

Catalog No. MP114004032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Powder; 1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114004032 Powder; 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114004032 Supplier MP Biomedicals™ Supplier No. 114004032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rich medium for the growth of S. cereviseae in liquid medium

  • Contains peptone and yeast extract
  • Useful for yeast requiring an alternative carbon source
  • Bulk powder
Formulation
  • Contents/liter: YPD, 17g agar
  • Use 67g per liter

Yeast Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Microbiology Media
Formulation Peptone 20g/L, Yeast extract-Y 10g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Quantity 1000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.