Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YEP Broth, Large Capsules
GSA_VA

Catalog No. MP114004046 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114004046 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114004046 Supplier MP Biomedicals™ Supplier No. 114004046
Add to Cart
Add to Cart

Rich medium for the growth of S. cereviseae in liquid medium

  • Contains peptone and yeast extract
  • Useful for yeast requiring an alternative carbon source
  • Pre-measured capsules
Formulation
  • Use 6 large capsules per liter
  • Contents/liter: 20g peptone, 10g yeast extract, pH 6.5 at 30°C

Yeast Cell Culture

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™ FDA
Content And Storage 15°C to 30°C
Format Capsule
Product Type Microbiology Media
Formulation Peptone 20g/L, Yeast extract-Y 10g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Quantity 5000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.