Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate

Catalog No. MP114027032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
454 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114027032 1000 g
MP114027022 454 g
MP114027012 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP114027032 Supplier MP Biomedicals™ Supplier No. 114027032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary

  • Nitrogen-based yeast

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Media Type YNB
Product Type Dehydrated Culture Media
Formulation Nitrogen Source: Ammonium Sulfate 5000mg/L; Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg; Trace Elements: Boric Acid 0.5mg, Copper Sulfate 0.04g, Potassium Iodide 0.1mg, Ferric Chloride 0.2mg, Manganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg
For Use With (Application) For vigorous growth of Saccharomyces cerevisiae
Quantity 1000 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.