Learn More
MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate/Copper Sulfate, Ferric Chloride
Shop All MP Biomedicals, Inc ProductsDescription
- Nitrogen-based yeast
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Specifications
| Certifications/Compliance | Molecular Biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Product Type | Microbiology Media |
| Formulation | Vitamins: Biotin 0.002mg/L, Calcium Pantothenate 0.4mg/L, Folic Acid 0.002mg/L, Inositol 2mg/L, Niacin 0.4mg/L, P-Aminobenzoic Acid 0.2mg/L, Pyridoxine Hydrochloride 0.4mg/L, Riboflavin 0.2mg/L, Thiamine Hydrochloride 0.4mg/L; Trace Elements: Boric Acid 0.5mg, Potassium Iodide 0.1mg, Maganese Sulfate 0.4mg, Sodium Molybdate 0.2mg, Zinc Sulfate 0.4mg; Salts: Magnesium Sulfate 500mg, Sodium Chloride 100mg, Potassium Phosphate monobasic 1000mg, Calcium Chloride 100mg |
| For Use With (Application) | For vigorous growth of Saccharomyces cerevisiae |
| Quantity | 227 g |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.