Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YNB, Powdered Media: Without Ammonium Sulfate/Folic Acid/Riboflavin
GSA_VA

Catalog No. MP114030512 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
227 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114030512 227 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114030512 Supplier MP Biomedicals™ Supplier No. 114030512
Add to Cart
Add to Cart

Can be prepared by adding a carbon source, the appropriate amino acids, and agar if necessary

  • Nitrogen-based yeast

Yeast cell culture, Gene expression, Transcription regulation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Microbiology Media
Formulation Trace Elements: Boric Acid 0.5mg/L, Copper Sulfate 0.04mg/L, Potassium Iodide 0.1mg/L, Ferric Chloride 0.2mg/L, Manganese Sulfate 0.4mg/L, Sodium Molybdate 0.2mg/L, Zinc Sulfate 0.4mg/L, Salts: Magnesium Sulfate 500mg/L, Sodium Chloride 100mg/L, Potassium Phosphate monobasic 1000mg, Calcium Chloride 132mg; Vitamins: 4-Amino Benzoic Acid 0.2mg, Biotin 0.002mg, Inositol 2mg, Nicotinic Acid 0.4mg, Panthotenic Acid Hemi Calcium Salt 0.4mg, Pyridoxine Hydrochloride 0.4mg, Thiamine Hydrochloride 0.4mg
For Use With (Application) For vigorous growth of Saccharomyces cerevisiae
Quantity 227 g
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.