Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YPD (YEPD) Broth
GSA_VA

Catalog No. MP114001075 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
10 x 1L pouch
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP114001075 10 x 1L pouch
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114001075 Supplier MP Biomedicals™ Supplier No. 114001075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rich medium for the growth of S. cerevisiae in liquid medium

  • Contains dextrose, peptone, and yeast extract
  • Includes ten 50g pouches
  • pH 6.5 at 30°C
Formulation
  • Add contents of one pouch to 1L of purified water
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose

Yeast Cell Culture

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Dehydrated
Product Type Microbiology Media
Formulation Peptone 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Packaging Type Pouch
Quantity 10 x 1000 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.