Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ YPD (YEPD) Broth, (Large Capsules)

Catalog No. mp114001036 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1000 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114001036 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP114001036 Supplier MP Biomedicals™ Supplier No. 114001036
Only null left
Add to Cart
Add to Cart

Rich medium for the growth of S. cereviseae in a liquid medium

  • Contains dextrose, peptone, and yeast extract
  • Pre-measured capsules
Formulation
  • Use 10 large capsules per liter
  • Contents/liter: 20g peptone, 10g yeast extract, 20g dextrose pH 6.5 at 30°C

Yeast Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™ FDA
Content And Storage 15°C to 30°C
Format Capsule
Product Type Microbiology Media
Formulation Peptone-Y 20g/L, Yeast extract 10g/L, Dextrose 20g/L
For Use With (Application) For growth of Saccharomyces cerevisiae in liquid medium
Quantity 1000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.