Learn More
Selleck Chemical LLC BRD0639-E1007-5MG
SDP
Supplier:  Selleck Chemical LLC E10075MG
Encompass_Preferred
BRD0639 is a first-in-class PBM-competitive small molecule inhibitor that disrupts the PRMT5-RIOK1 complex with IC50s of 7 5 M and 16 M in permeabilized and living cells respectively
Catalog No. 50-313-7244
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.