Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP1/MAP2K1IP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NBP150631H
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP150631H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP150631H Supplier R&D Systems Supplier No. NBP150631H

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

MP1/MAP2K1IP1 Polyclonal specifically detects MP1/MAP2K1IP1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MP1/MAP2K1IP1
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Accession No. Q9UHA4
Gene Alias Late endosomal/lysosomal adaptor and MAPK and MTOR activator 3, late endosomal/lysosomal adaptor, MAPK and MTOR activator 3, MAP2K1IP1, MAPBP, MAPK scaffold protein 1, MAPKSP1, MEK binding partner 1, MEK partner 1, MEK-binding partner 1, mitogen-activated protein kinase kinase 1 interacting protein 1, Mitogen-activated protein kinase kinase 1-interacting protein 1, Mitogen-activated protein kinase scaffold protein 1, Mp1, PRO0633, Ragulator3
Gene Symbols LAMTOR3
Host Species Rabbit
Immunogen A genomic peptide made to an internal region of the human MAP2K1IP1/MAPKSP1 protein (within residues 20-180). [Swiss-Prot Q9UHA4]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8649
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.