Learn More
Perkin Elmer US LLC MEINHARD Nebulizer Gas Line
Supplier:  Perkin Elmer US LLC N0791843
Encompass_Preferred
This gas line is designed for use with MEINHARD Concentric Nebulizers. The MEINHARD Concentric Nebulizer reflects exacting standards in design and manufacturing. They provide excellent sensitivity and precision for aqueous solutions and samples with few dissolved solids (less than 1%). MEINHARD Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid.
Catalog No. 50-211-2345
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.