Learn More
Invitrogen™ MP68 Polyclonal Antibody

Description
Immunogen sequence: PMKPYYTKVY QEIWIGMGLM GFIVYKIRAA DKRSKALKAS APAPGHH Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 76%.
Specifications
Specifications
| Antigen | MP68 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Atp5mpl |
| Gene Accession No. | P56378 |
| Gene Alias | 2010107E04Rik; 6.8 kDa mitochondrial proteolipid; 6.8 kDa mitochondrial proteolipid protein; 9430003J03Rik; ATP synthase membrane subunit 6.8PL; ATP synthase subunit ATP5MPL, mitochondrial; Atp5mpl; AU043134; AV124504; C14orf2; chromosome 14 open reading frame 2; MLQ; Mp68; PLPM; PRO1574; RIKEN cDNA 2010107E04 gene |
| Gene Symbols | Atp5mpl |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.