Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ mPAGE™ Color Protein Standard

Catalog No. MPSTD4MI Shop All MilliporeSigma Products
Change view
Click to view available options
mPAGE™ Color Protein Standard
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MPSTD4MI mPAGE™ Color Protein Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MPSTD4MI Supplier MilliporeSigma™ Supplier No. MPSTD4
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

SDS-PAGE, western blot, Color protein standard

  • Mixture of 10 highly purified prestained proteins ranging from 10 kDa to 203 kDa, covalently coupled with different chromophores
  • Designed for observing protein separation during SDS-PAGE, verifying western transfer efficiency on membranes, and approximating the size of proteins

Thaw and mix at room temp before use, only suitable for rough estimation of protein molecular weight

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 10 to 203 kDa
Content And Storage -20°C for long term storage, 4°C for up to 3 months or 25°C for up to two weeks
Cell Line 25 to 10 μL/Lane
Buffer 62.5 mM Tris-H3PO4 (pH 7.5 (at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 33 & glycerol)

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.