Learn More
MilliporeSigma mPAGE™ Mini Wet Transfer System
Catalog No. 11580588 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
Protein Transfer
Catalog No. For Use With (Application)
11-580-588 Protein Transfer
1 options
Catalog No. 11-580-588 Supplier MilliporeSigma Supplier No. MWTS
Add to Cart
Add to Cart
System for transfer of proteins from a mini acrylamide gel to blotting membrane, accommodates two wet transfer cassettes
  • The mPAGE™ Mini Wet Transfer System accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE™ Transfer Buffer or a compatible transfer buffer.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Protein Transfer
Type Wet Transfer System