Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ mPAGE™ Unstained Protein Standard

Catalog No. MPSTD3MI Shop All MilliporeSigma Products
Change view
Click to view available options
Sample Volume:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
MPSTD3MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPSTD3MI Supplier MilliporeSigma™ Supplier No. MPSTD3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SDS-PAGE, western blot

  • Mixture 12 recombinant unstained proteins 10 kDA to 200 kDa
  • Higher intensity bands: 85 kDa, 25 kDa,10 kDa (for reference)
  • Suitable for molecular weight determination after SDS-PAGE or Western blotting
  • Serves as standard to calibrate mobility of prestained markers
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 10 to 200 kDa
Sample Volume 500 μL
Content And Storage -20°C; for up to one yr, 4°C for up to three months or 25°C; up to two weeks
Buffer 20 mM Tris-phosphate)pH 7.5 (at 25°C), 2% SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, 15% Glycerol)

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.