Learn More
Sigma Aldrich Fine Chemicals Biosciences mPEG40K MAL average Mn 4001G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences JKA31231G
Encompass
Methoxy PEG Maleimide. Selective for thiol groups on cystein side chains reacts at pH 5.0-6.6
Catalog No. 50-288-6462
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.