Learn More
Enzo Life Sciences Myeloperoxidase (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900115
Encompass_Preferred
The Myeloperoxidase (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Tihs sensitive kit lets you quantitate as little as 19 pg/mL MPO in a wide variety of sample matrices that have been validated in-house. Convenient ready-to-use liquid color-coded reagents reduce error and save time in the lab. Alternative Name: MPO. Sensitivity: 0.019 ng/ml (range 0.195 - 12.5 ng/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 13, Assay buffer 31 concentrate, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2
Catalog No. 50-658-151