Learn More
Description
Includes
- MPO Microplate (Item A): 96 wells coated with anti-mouse MPO
- Wash Buffer Concentrate, 20x(Item B): 20x concentrated solution
- Standards (Item C): Mouse MPO (1 vial is enough to run each standard in duplicate)
- Detection Antibody (Item F): Biotinylated anti-mouse MPO
- HRP-Streptavidin Concentrate (Item G): 400x concentrated HRP-conjugated streptavidin
- TMB One-Step Substrate Reagent (Item H): 3,3',5,5'-tetramethylbenzidine (TMB) in,buffer solution
- Stop Solution (Item I): 0.2 M sulfuric acid
- Assay Diluent C (Item L): Diluent buffer
- Assay Diluent B (Item E): 5x concentrated buffer
Specifications
Specifications
| Assay Sensitivity | 600 pg/mL |
| Content And Storage | Store the kit at 4°C for short term storage. Rconstituted standard should be stored at -80°C. This kit can be stored at -20°C for long term storage. Avoid repeated freezing and thawing. |
| Description | Mpo (Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of mouse Mpo. |
| Conjugate | HRP (Horseradish Peroxidase) |
| Cross Reactivity | Mouse |
| Detection Method | Colorimetric |
| Product Type | Mpo (Mouse) ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma |
| Sufficient For | 96 |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.