Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MPO (Mouse) ELISA Kit

Catalog No. 89101102 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP (Horseradish Peroxidase)
Sample Type:
Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Cell Culture Supernatant, Plasma
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate Sample Type
89-101-102 Biotin Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
89-100-459 HRP (Horseradish Peroxidase) Cell Culture Supernatant, Plasma
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-102 Supplier Abnova™ Supplier No. KA3690
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-mouse MPO antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse MPO standard, 10ng x 2
  • Biotinylated anti-mouse MPO antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Mpo. Sandwich enzyme immunoassay.

Mpo (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 156 to 10000pg/mL
Content And Storage Store the kit at -20°C
Description Mpo (Mouse) is a sandwich enzyme immunoassay for the quantitative measurement of mouse Mpo.
Conjugate Biotin
Cross Reactivity Mouse
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and recombinant mouse MPO
Sufficient For 96
For Use With (Application) Quantification
Kit Contents 96-well plate precoated with anti- mouse MPO antibody (96 (8 x 12) wells), Lyophilized recombinant mouse MPO standard (10ng x 2), Biotinylated anti- mouse MPO antibody (dilution 1:100): 13μL, Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100): 130μL, Sample diluent buffer (30mL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10 pg/mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.