Learn More
CAPRICO BIOTECHNOLOGIES INC MPO PerCP
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4123035
Encompass_Preferred
The Caprico MPO PerCP antibody is a mouse monoclonal antibody that recognizes myeloperoxidase, an enzyme expressed in myeloid cells including neutrophils, monocytes, and some myeloid progenitor cells. This antibody is conjugated to PerCP, a fluorescent dye, allowing detection of MPO expression by flow cytometry. The antibody is supplied as a liquid in PBS with 0.1% sodium azide and 1% BSA. Each vial contains 100 tests of antibody at a concentration of 0.2 mg/mL. The antibody is recommended for flow cytometry applications. Proper handling, storage, and dilution guidelines are provided. This MPO antibody enables identification and analysis of myeloid cell populations.
Catalog No. 50-227-9318
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.