Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MPP dihydrochloride
GSA_VAGREENER_CHOICE

Catalog No. 199110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-911-0 10 mg
19-915-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-911-0 Supplier Tocris Bioscience™ Supplier No. 1991/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly selective ERα antagonist

Selective, high affinity silent antagonist at ERα receptors. Displays > 200-fold selectivity for ERα over ERβ. Ki values are 2.7 and 1800 nM at ERα and ERβ receptors respectively.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 911295-24-4
Molecular Formula C29H33Cl2N3O3

Specifications

Quantity 10 mg
Formula Weight 542.5
Purity >98%
Molecular Weight (g/mol) 542.501
Chemical Name or Material MPP dihydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.