Learn More
Invitrogen™ MPP10 Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 79 kDa and 120 kDa (phosphorylated). Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | MPP10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MPHOSPH10 |
| Gene Accession No. | O00566, Q810V0 |
| Gene Alias | 2810453H10Rik; 5730405D16Rik; AI326008; CT90; M phase phosphoprotein 10; M-phase phosphoprotein 10; M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein); Mphosph10; MPP10; MPP10P; PPP1R106; protein phosphatase 1, regulatory subunit 106; U3 small nucleolar ribonucleoprotein; U3 small nucleolar ribonucleoprotein protein MPP10 |
| Gene Symbols | MPHOSPH10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.