Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP3 Monoclonal Antibody (OTI2A11), TrueMAB™, OriGene
SDP

Catalog No. p-200009805
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2185 30 μL
50-167-2701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2185 Supplier OriGene Supplier No. TA501564S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MPP3 Monoclonal antibody specifically detects MPP3 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen MPP3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2A11
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Prpsap2
Gene Accession No. O60256
Gene Alias 6430514B01; CSG18; discs large homolog 3; discs, large homolog 3; DLG3; Dlgh3; Dusp3; MAGUK p55 subfamily member 3; MAGUK p55 subfamily member 3 {ECO:0000250; membrane palmitoylated protein 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Mpp3; mpp3 {ECO:0000312; Mpp3 membrane protein, palmitoylated 3; protein Dlgh3; protein MPP3; RGD:620015}; UniProtKB:O88910}
Host Species Mouse
Immunogen Full length human recombinant protein of human PRPSAP2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5636
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.