Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PureAire Monitoring Systems MPS Flammable Gas Monitor-Ethanol 0-100 LEL
SDP

Catalog No. 17542404
Change view
Click to view available options
Components Measured:
Ethanol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-542-404 Ethanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-542-404 Supplier PureAire Monitoring Systems Supplier No. 98005
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PureAire’s MPS LEL Ethanol Gas Monitor offers accurate, continuous monitoring in hazardous areas. The durable, poison-resistant MPS sensor lasts 15+ years, requires no calibration, and auto-adjusts for temperature, humidity, and pressure. A magnetic keypad allows full access without opening the case.

  • Detects Ethanol 0-100% LEL
  • 15+ Year sensor - Zero calibration required
  • Mean Time To Failure (Sensor Only): 62 years at 60% confidence or 19 years at 99% confidence
  • RS-485 Communication Interface
  • Easy to use - Set it and forget it technology
  • Zero drift
  • Immune to poisoning
  • Operating temperatures: -4°F to +140°F (-20°C to +60°C)
  • Fail-safe: Built-in self-test
  • Back-lit LCD Display
  • 4-20mA output
  • User-adjustable dual alarm relays
  • Durable XP Housing: Certified for hazardous locations
  • Ratings: Class I and II, Division 1, Groups B, C, D, E, F, G. Class III Type 4X/IP66 ∣ ATEX* - Ex d IIC ∣ Ex d IIC IEC 60529 IP66 ∣ IECEx* Ex db IIC T4 Gb ∣ Tamb - 20°C to +60°C

Specifications

Components Measured Ethanol
For Use With (Equipment) Gas Detector
Type Fixed Combustible/Toxic Gas Detector
IP Rating IP66
Voltage 21 to 24 VDC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.