Learn More
AdipoGen MPTS
SDP
Supplier:  AdipoGen CDXM0055G001
Encompass_Preferred
Chemical. CAS 82962-86-5. Formula C17H9Na3O10S3. MW 538.41. Synthetic. Highly water-soluble superpolar fluorescent probe for assaying cationic surfactants. Can be used as a polar tracer. It has also been used in combination with a boronic acid-modified viologen quencher to sense glucose at pH 7.4 in buffered aqueous solution.
Catalog No. 50-256-0551
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.