Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPZL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984137
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4137 Supplier Bioss Supplier No. BS17744R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cell surface receptor, which is involved in signal transduction processes. Recruits PTPN11/SHP-2 to the cell membrane and is a putative substrate of PTPN11/SHP-2. Is a major receptor for concanavalin-A (ConA) and is involved in cellular signaling induced by ConA, which probably includes Src family tyrosine-protein kinases. Isoform 3 seems to have a dominant negative role; it blocks tyrosine phosphorylation of MPZL1 induced by ConA. Isoform 1, but not isoform 2 and isoform 3, may be involved in regulation of integrin-mediated cell motility.

Specifications

Antigen MPZL
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mpzl1
Gene Accession No. O95297
Gene Alias 1110007A10Rik; FLJ21047; immunoglobulin family transmembrane protein; MPZL1; MPZL1b; myelin protein zero like 1; myelin protein zero-like 1; Myelin protein zero-like protein 1; protein zero related; Protein zero-related; Pzr; PZR1b; PZRa; PZRb; UNQ849/PRO1787
Gene Symbols Mpzl1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MPZL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9019
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.