Learn More
AdipoGen MQAE
SDP
Supplier:  AdipoGen CDXE0001G001
Encompass_Preferred
Chemical. CAS 162558-52-3. Formula C14H16NO3Br. MW 326.19. Synthetic. Improved chloride probe. Used to measure chloride transport in liposome membranes.
Catalog No. 50-256-1005
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.