Learn More
Description
Includes
Tube containing 100 strips
- Small and easy to handle
- Flexible and universal application
- EDTA reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
1.10025.0001
Color-scale graduation: 4 - 5 - 9 - 18 - 26°e
1.10046.0001
Color-scale graduation: 6 - 13 - 19 - 25 - 31°e
Reclose the tube containing the test strips immediately after use.
Analytical Quality Assurance, Standard for Total Hardness
Specifications
Specifications
| For Use With (Application) | Analytical quality assurance, to determine the total hardness of water |
| For Use With (Equipment) | Colorimeter |
| Graduations | 4 to 26°e |
| No. of Tests | 5000 |
| Range | 4 to 26°e |
| Storage Requirements | 15°C to 25°C |
| Test Method | EDTA |
| Type | Test Strip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.