Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552775 Supplier Invitrogen™ Supplier No. PA552775
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MANGTNASAP YYSYEYYLDY LDLIPVDEKK LKAHKHS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

MRAP encodes a melanocortin receptor-interacting protein. The encoded protein regulates trafficking and function of the melanocortin 2 receptor in the adrenal gland. The encoded protein can also modulate signaling of other melanocortin receptors. Mutations in this gene have been associated with familial glucocorticoid deficiency type 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRAP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mrap
Gene Accession No. Q8TCY5
Gene Alias 1110025G12Rik; B27; C21orf61; Falp; Fat cell-specific low molecular weight protein; fat tissue-specific low MW protein; FGD2; GCCD2; melanocortin 2 receptor accessory protein; melanocortin-2 receptor accessory protein; Mrap; ORF61
Gene Symbols Mrap
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRAP. Recombinant protein control fragment (Product #RP-90458).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56246
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.