Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRAS Recombinant Mouse Monoclonal Antibody (17H12)
GREENER_CHOICE

Catalog No. FEMA550225
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550225 Supplier Invitrogen™ Supplier No. MA550225
Only null left
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

MRAS Recombinant Monoclonal Antibody for ICC/IF, IHC (P), ELISA

MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRAS
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 17H12
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MRAS
Gene Accession No. O14807
Gene Alias 2900078C09Rik; AI326250; MRAS; M-RAs; Muscle and microspikes Ras; muscle RAS oncogene homolog; ras-related protein M-Ras; Ras-related protein R-Ras3; RRAS3; R-RAS3; Xras; X-Ras
Gene Symbols MRAS
Host Species Mouse
Immunogen Recombinant Human MRAS protein.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22808
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.