Learn More
Cygnus Technologies MRC5 HCP ELISA Kit
Supplier:  Cygnus Technologies F300
Encompass
The MRC5 (human fetal lung fibroblast) cell line is a cost-effective method for the production of vaccines and supports the growth of a broad range of viruses. Our assay qualification has determined that the LOD for this kit is ~1.8 ng/ml.
Catalog No. NC1680849
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.