Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRCK alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584863 Supplier Invitrogen™ Supplier No. PA584863
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TRRESQSERE EFESEFKQQY EREKVLLTEE NKKLTSELDK LTTLYENLSI HNQQLEEEVK D Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

CDC42BPA (MRCKA) is a serine/threonine kinase which functions in actin cytoskeleton assembly and organization. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRCK alpha
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDC42BPA
Gene Accession No. Q5VT25
Gene Alias A930014J19Rik; CDC42 binding protein kinase alpha; CDC42 binding protein kinase alpha (DMPK-like); CDC42 binidng protein kinase beta; CDC42-binding protein kinase alpha; CDC42-binding protein kinase alpha (DMPK-like); CDC42BPA; cdc42bpa {ECO:0000250; DKFZp686L1738; DKFZp686P1738; DMPK-like; DMPK-like alpha; FLJ23347; Kiaa0451; MRCK; MRCK alpha; MRCKA; myotonic dystrophy kinase-related CDC42-binding kinase alpha; myotonic dystrophy kinase-related CDC-42-binding kinase-alpha; myotonic dystrophy kinase-related CDC42-binding protein kinase alpha; myotonic dystrophy protein kinase-like alpha; mytonic dystrophy kinase-related Cdc42-binding kinase; PK428; RP5-1087E8.4; serine/threonine-protein kinase MRCK alpha; ser-thr protein kinase PK428; ser-thr protein kinase related to the myotonic dystrophy protein kinase; UniProtKB:Q5VT25}
Gene Symbols CDC42BPA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDC42BPA. Recombinant protein control fragment (Product #RP-107808).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8476
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.