Learn More
Description
Suggested positive control: 293 whole cell lysate.
Specifications
Specifications
| Antigen | MRE11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Mre11 |
| Gene Accession No. | P49959 |
| Gene Alias | ATLD; AT-like disease; DNA recombination and repair protein; double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; HNGS1; Meiotic recombination 11 homolog 1; meiotic recombination 11 homolog A; meiotic recombination 11 homolog A (S. cerevisiae); mmMRE11A; MRE11; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog A, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11 homolog A (S. cerevisiae); MRE11 meiotic recombination 11-like protein A; MRE11A; MRE11B; MRX complex nuclease subunit; NGS1; RAD58; XRS4; YM9959.06C; YMR224C |
| Gene Symbols | Mre11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
