Learn More
Description
Includes
- 1.NB100-142: Mre11 Antibody
- 2.NB100-143: Nbs1 Antibody
- 3.NB100-154: Rad50 Antibody
Specifications
Specifications
| Antigen | Mre11-Rad50-NBS1 Complex |
| Applications | ChIP Assay, ELISA, Flow Cytometry, Gene Knock-Out, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Chromatin Immunoprecipitation (ChIP), Knockout Validated, Knockdown Validated |
| Gene Alias | ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11MRE11B |
| Gene Symbols | MRE11 |
| Host Species | Rabbit |
| Immunogen | See individual datasheets. |
| Quantity | 3 Vials |
| Gene ID (Entrez) | 4361 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
