Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mreB Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144554 Supplier Invitrogen™ Supplier No. PA5144554
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mreB Polyclonal Antibody for Western Blot, ELISA

Forms membrane-associated dynamic filaments that are essential for cell shape determination (PubMed:15612918, PubMed:21903929). Acts by regulating cell wall synthesis and cell elongation, and thus cell shape (PubMed:21903929). A feedback loop between cell geometry and MreB localization maintains elongated cell shape by targeting cell wall growth to regions of negative cell wall curvature (PubMed:24550515). Filaments rotate around the cell circumference in concert with the cell wall synthesis enzymes. The process is driven by the cell wall synthesis machinery and does not depend on MreB polymerization (PubMed:21903929). Rotation may contribute to the robust maintenance of rod shape (PubMed:21903929).
TRUSTED_SUSTAINABILITY

Specifications

Antigen mreB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene mreB
Gene Accession No. P0A9X4
Gene Alias envB; rodY
Gene Symbols mreB
Host Species Rabbit
Immunogen Recombinant Escherichia coli (strain K12) mreB protein (1-347aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 82262299
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.