Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGPRX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113198
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113198 Supplier Invitrogen™ Supplier No. PA5113198
Only null left
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

MRGPRX2 Polyclonal Antibody for ELISA, ICC/IF, Western Blot

Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Cortistatin-14 seems to be a high potency ligand at this receptor. Cortistatin has several biological functions including roles in sleep regulation locomotor activity, and cortical function. In receptor-expressing cells, cortistatin-stimulated increases in intracellular Ca(2+) but had no effect on basal or forskolin-stimulated cAMP levels, suggesting that this receptor is G(q)-coupled. Has a limited expression profile, both peripheral and within the central nervous system, with highest levels in dorsal root ganglion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGPRX2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MRGPRX2
Gene Accession No. Q3UG50, Q96LB1
Gene Alias G protein-coupled receptor MRGX2; G370024M05Rik; MAS related GPR family member X2; Mas-related G protein-coupled receptor G3; MAS-related GPR, member B10; MAS-related GPR, member X2; mas-related G-protein coupled receptor member B10; Mas-related G-protein coupled receptor member X2; MGRG3; MrgB10; Mrgprb10; Mrgprx2; MRGX2
Gene Symbols MRGPRX2
Host Species Rabbit
Immunogen Recombinant Human Mas-related G-protein coupled receptor member X2 protein (286-330AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117194, 243978
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.