Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRGPRX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533952
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533952 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533952 Supplier Invitrogen™ Supplier No. PA533952
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Gorilla (94%) Monkey (82%).

Orphan receptor activated by a subset of RFamide-family neuropeptides such as FLRF-amide and FMRF-amide. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system. Its effect is mediated by G(q) and G(11) proteins. May regulate the function of nociceptive neurons by modulation of pain perception. Expressed in a subset of IB4-positive small diameter nociceptive dorsal root neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGPRX3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MRGPRX3
Gene Accession No. Q96LB0
Gene Alias adenine receptor; G protein-coupled receptor MRGX3; G protein-coupled receptor SNSR1; G protein-coupled receptor SNSR2; GPCR; G-protein coupled receptor; G-protein coupled receptor; rc 56.1.3; Hdrr1; HDRR2; MAS related GPR family member X3; Mas-related G protein-coupled receptor g1; Mas-related gene A; mas-related GPCR member X3; MAS-related GPR, member X3; mas-related G-protein coupled receptor member A; Mas-related G-protein coupled receptor member X3; MAS-related G-protein coupled receptor, member A; MGC108580; Mgrg1; Mrga; Mrga10; Mrgpra; MRGPRX3; MRGX3; nuclear receptor MrgA10 RF-amide G protein-coupled receptor; rc 56.1.3; sensory neuron-specific G-protein coupled receptor 1/2; SNSR1; SNSR2
Gene Symbols MRGPRX3
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 1st extracellular domain of human MRGPRX3 / MRGX3.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117195
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.