Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518973
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518973 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518973 Supplier Invitrogen™ Supplier No. PA518973
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra-and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This full transporter is a member of the MRP subfamily which is involved in multi-drug resistance. This protein functions as a multispecific organic anion transporter, with oxidized glutathione, cysteinyl leukotrienes, and activated aflatoxin B1 as substrates. This protein also transports glucuronides and sulfate conjugates of steroid hormones and bile salts. Alternative splicing by exon deletion results in several splice variants but maintains the original open reading frame in all forms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ABCC1
Gene Accession No. P33527
Gene Alias ABC29; ABCC; Abcc1; Abcc1a; Abcc1b; ATP binding cassette subfamily C member 1; ATP-binding cassette sub-family C ( CFTR/MRP) member 1a; ATP-binding cassette sub-family C member 1; ATP-binding cassette transporter variant ABCC1delta-ex13; ATP-binding cassette transporter variant ABCC1delta-ex13&14;; ATP-binding cassette transporter variant ABCC1delta-ex25; ATP-binding cassette transporter variant ABCC1delta-ex25&26; ATP-binding cassette, subfamily C (CFTR/MRP), member 1; ATP-binding cassette, sub-family C (CFTR/MRP), member 1; ATP-binding cassette, sub-family C (CFTR/MRP), member 1a; ATP-binding cassette, sub-family C (CFTR/MRP), member 1b; Avcc1a; DKFZp686N04233; DKFZp781G125; Glutathione-S-conjugate-translocating ATPase ABCC1; GS-X; leuk; leukotriene C(4) transporter; LTC4 transporter; Mdrap; MRP; Mrp1; multidrug resistance protein 1; multidrug resistance-associated protein 1; multiple drug resistance-associated protein
Gene Symbols ABCC1
Host Species Goat
Immunogen Synthetic peptide sequence (HQSDLKVDENQKAYY) corresponding to the internal amino acids of ABCC1 (aa 1176-1191).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4363
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.