Learn More
Invitrogen™ MRP5 Monoclonal Antibody (M5I-1)

Description
M5I-1 reacts with an internal epitope of MRP5, a 190-200 kD transmembrane protein that is closely related to the multidrug resistance protein MRP. M5I-1 was raised against a bacterial fusion protein of MRP5, containing amino acids 82-168 of the protein. M5I-1 does not cross-react with the human MDR1, MRP1, MRP2 or MRP3 gene products.
Specifications
Specifications
| Antigen | MRP5 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M5I-1 |
| Concentration | 250 μg/mL |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.7% BSA and 0.1% sodium azide |
| Gene | ABCC5 |
| Gene Accession No. | O15440 |
| Gene Alias | 2900011L11Rik; ABC 33; ABC33; ABCC 5; ABCC5; Abcc5a; Abcc5b; AI132311; ATP binding cassette subfamily C member 5; ATP-binding cassette sub-family C (CFTR/MRP) member 5a; ATP-binding cassette sub-family C member 5; ATP-binding cassette, subfamily C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5a; ATP-binding cassette, sub-family C (CFTR/MRP), member 5b; ATP-binding cassette, sub-family C, member 5; canalicular multispecific organic anion transporter C; DKFZp686C1782; EST277145; MOAT C; MOATC; MOAT-C; MRP 5; MRP5; multidrug resistance-associated protein 5; Multi-specific organic anion transporter C; pABC 11; pABC11; SMRP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.