Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597980
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597980 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597980 Supplier Invitrogen™ Supplier No. PA597980
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L3P ribosomal protein family. A pseudogene corresponding to this gene is found on chromosome 13q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MRPL3
Gene Accession No. P09001
Gene Alias 2010320L16Rik; 39S ribosomal protein L3, mitochondrial; 5930422H18Rik; AA407889; COXPD9; L3mt; mitochondrial 60S ribosomal protein L3; Mitochondrial large ribosomal subunit protein uL3m; mitochondrial ribosomal protein L3; MRL3; Mrpl3; MRP-L3; RPML3
Gene Symbols MRPL3
Host Species Rabbit
Immunogen Recombinant Human 39S ribosomal protein L3, mitochondrial protein (201-348AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11222
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.