Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL51 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558988 Supplier Invitrogen™ Supplier No. PA558988
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GNFEKHPKEL IRGPIWLRGW KGNELQRCIR KRKMVGSRMF ADDLHNLNKR IRYLYKHFNR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.

Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-L51(mitochondrial ribosomal protein L51), also known as CDA09, MRP64, bMRP64 or HSPC241, is a 128 amino acid mitochondrial ribosomal protein. MRP-L51 is a component of the mitochondrial ribosome large subunit (39S) which consists of a 16S rRNA and about 50 distinct proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL51
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL51
Gene Accession No. Q4U2R6
Gene Alias 2610511M02Rik; 39S ribosomal protein L51, mitochondrial; bMRP64; bMRP-64; CDA09; HSPC241; L51mt; Mitochondrial large ribosomal subunit protein mL51; mitochondrial ribosomal protein 64; mitochondrial ribosomal protein bMRP64; mitochondrial ribosomal protein L51; MRP64; MRPL51; MRP-L51; rm51
Gene Symbols MRPL51
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL51. Recombinant protein control fragment (Product #RP-97689).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.