Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPL55 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597981
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597981 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597981 Supplier Invitrogen™ Supplier No. PA597981
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRPL55 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL55 encodes a 39S subunit protein. Multiple transcript variants encoding two different isoforms were identified through sequence analysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL55
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MRPL55
Gene Accession No. Q7Z7F7
Gene Alias 2810038N09Rik; 39S ribosomal protein L55, mitochondrial; AAVG5835; L55mt; L55nt; Mitochondrial large ribosomal subunit protein bL31m; Mitochondrial large ribosomal subunit protein mL55; mitochondrial ribosomal protein L55; Mrpl55; MRP-L55; PRO19675; Unknown (protein for MGC:137378); UNQ5835/PRO19675
Gene Symbols MRPL55
Host Species Rabbit
Immunogen Recombinant Human 39S ribosomal protein L55, mitochondrial protein (34-128AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128308
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.