Learn More
Aviva Systems Biology Corp MRPS18B Antibody - N-terminal region (ARP62873_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62873_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MRPS18B. It was validated on Western Blot. . Alternate names: C6orf14, DKFZp564H0223, HSPC183, HumanS18a, MRP-S18-2, MRPS18-2, PTD017, S18amtApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-795
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.