Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139985 Supplier Invitrogen™ Supplier No. PA5139985
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PVCAKNRAAR VRVSKGDKPV TYEEAHAPHY IAHRKGWLSL HTGNLDGEDH AAERTVEDVF LRKFMW Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS24 encodes a 28S subunit protein. A pseudogene corresponding to this gene is found on chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS24
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS24
Gene Accession No. Q96EL2
Gene Alias 28S ribosomal protein S24, mitochondrial; 3110030K20Rik; AI414579; bMRP47; bMRP-47; DD6A4-3; HSPC335; mitochondrial 28S ribosomal protein S24; mitochondrial ribosomal protein S24; Mitochondrial small ribosomal subunit protein uS3m; MRPS24; MRP-S24; ribosomal protein, mitochondrial, S24; Rpms24; S24mt
Gene Symbols MRPS24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS24. Recombinant protein control fragment (Product #RP-109377).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64951
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.