Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553844
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553844 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553844 Supplier Invitrogen™ Supplier No. PA553844
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPEPEPALDL AALRAVACDC LLQEHFYLRR RRRVHRYEES EVISLPFLDQ LVSTLVGLLS PHNPALAAAA LDYRCPVHFY WVRGEEIIPR GH Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 53%.

Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-S30 (mitochondrial ribosomal protein S30), also known as PDCD9 (programmed cell death protein 9), is a 439 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. MRP-S30 is expressed in kidney, liver, heart and skeletal muscle. The gene encoding MRP-S30 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS30
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mrps30
Gene Accession No. Q9NP92
Gene Alias 2610020A16Rik; 28S ribosomal protein S30, mitochondrial; 39S ribosomal protein S30, mitochondrial; AA968347; BM-047; Mitochondrial large ribosomal subunit protein mL65; Mitochondrial large ribosomal subunit protein mS30; mitochondrial ribosomal protein S30; MRP S30; Mrps30; MRP-S30; PAP; PDCD9; programmed cell death 9; Programmed cell death protein 9; S30mt
Gene Symbols Mrps30
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS30. Recombinant protein control fragment (Product #RP-92027).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.