Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563173 Supplier Invitrogen™ Supplier No. PA563173
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QETHQQLADK KGLHVVEIRE ECGPLPIVVA SPRGPLRKDP EPEDEVPDVK LDWEDVKTAQ GMKRSVWSNL K Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.

Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-S5 (mitochondrial ribosomal protein S5), alternatively known as S5mt, is a 430 amino acid mitochondrial protein belonging to the ribosomal protein S5P family. A component of the 28S subunit, MRP-S5 contains one S5 DRBM domain and exists as two alternatively spliced isoforms. The gene encoding MRP-S5 maps to human chromosome 2, which consists of 237 million bases, encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2 including Harlequin icthyosis, sitosterolemia and Alstrom syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPS5
Gene Accession No. P82675
Gene Alias 1620401I16Rik; 28S ribosomal protein S5, mitochondrial; AI850294; mitochondrial 28S ribosomal protein S5; mitochondrial ribosomal protein S5; Mitochondrial small ribosomal subunit protein uS5m; MRPS5; MRP-S5; S5mt
Gene Symbols MRPS5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPS5. Recombinant protein control fragment (Product #RP-101798).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.