Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MRPS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588239 Supplier Invitrogen™ Supplier No. PA588239
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEEFVQRFRR SVTLESKKQL IEPVQYDEQG MAFSKSEGKR KTAKAEAIVY KHGSGRIKVN GIDYQLYFPI TQDREQLMFP FHFVDRLGKH DVTCTVSGGG RSAQAGAIRL AMAKALCSFV TEDEVEWMRQ AGLLTTDPRV RERKKPGQEG ARRKFTWKKR; Positive Samples: LO2, U-87MG, A-431, HT-29; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Pseudogenes corresponding to this gene are found on chromosomes 2p and 17q.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPS9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPS9
Gene Accession No. P82933
Gene Alias 2310002A08Rik; 28S ribosomal protein S9, mitochondrial; AW322516; mg637; mitochondrial ribosomal protein S9; Mitochondrial small ribosomal subunit protein uS9m; MRPS9; MRP-S9; muscle protein 637; RPMS9; S9mt
Gene Symbols MRPS9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 237-396 of human MRPS9 (NP_8725781).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64965
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.