Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRS 1845, Tocris Bioscience™

Catalog No. 186650
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-665-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-665-0 Supplier R&D Systems Supplier No. 1866/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Potent SOCE inhibitor; blocks capacitative Ca2+ entry

Potent blocker of store-operated Ca 2+ channels; inhibits capacitative Ca 2+ influx in HL-60 cells (IC 50 = 1.7 mM).

Chemical Identifiers

CAS 544478-19-5
Molecular Formula C21H22N2O6
Molecular Weight (g/mol) 398.415
InChI Key BITHABUTZRAUGT-UHFFFAOYSA-N
Synonym n-propargylnitrendipene, n-propylargylnitrendipine, mrs hplc , solid, 1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-1-2-propynyl-3,5-pyridinedicarboxylic acid ethyl, methyl ester, 3-ethyl 5-methyl 2,6-dimethyl-4-3-nitrophenyl-1-prop-2-yn-1-yl-4h-pyridine-3,5-dicarboxylate
PubChem CID 11538542
IUPAC Name 5-O-ethyl 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1-prop-2-ynyl-4H-pyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(N(C(=C(C1C2=CC(=CC=C2)[N+](=O)[O-])C(=O)OC)C)CC#C)C

Specifications

Quantity 50 mg
Formula Weight 398.41
Percent Purity >98%
Chemical Name or Material MRS 1845
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.