Learn More
Selleck Chemical LLC MRS 2578 10mg 711019-86-2
SDP
Supplier:  Selleck Chemical LLC S285510MG
Encompass_Preferred
MRS2578 is a potent P2Y6 receptor antagonist with IC50 of 37 nM, exhibits insignificant activity at P2Y1, P2Y2, P2Y4,and P2Y11 receptors. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3066
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.